(5-Fluoro-2,3-dihydrobenzo[b][1,4]dioxin-6-yl)boronic acid ≥97%

Supplier: AOBChem USA

Warning

20614-5G 20614-1G 20614-25G 20614-500MG
77628-076EA 1818.98 USD
77628-076 77637-168 77654-136 77645-176
(5-Fluoro-2,3-dihydrobenzo[b][1,4]dioxin-6-yl)boronic acid ≥97%
(5-Fluoro-2,3-dihydrobenzo[b][1,4]dioxin-6-yl)boronic acid

Precaución: For Research Use Only

Formula: 197.956 g/mol
MW: C₈H₈BFO₄
Storage Temperature: Ambient
MDL Number: MFCD23380875
CAS Number: 1313426-14-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR