2,5-Dibromo-1-(methoxymethoxy)benzene ≥95%

Supplier: AOBChem USA

Warning

20604-5G 20604-25G
77634-338EA 176 USD
77634-338 77654-132
2,5-Dibromo-1-(methoxymethoxy)benzene ≥95%
2,5-Dibromo-1-(methoxymethoxy)benzene

Precaución: For Research Use Only

Formula: C₈H₈Br₂O₂
Storage Temperature: Ambient
MDL Number: MFCD31805743
CAS Number: 2271442-85-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR