4-(Methoxymethyl)-2,6-dimethylphenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

20541-5G 20541-25G 20541-1G 20541-500MG
77631-982EA 1380 USD
77631-982 77657-942 77641-184 77649-228
4-(Methoxymethyl)-2,6-dimethylphenylboronic acid ≥97%
4-(Methoxymethyl)-2,6-dimethylphenylboronic acid

Precaución: For Research Use Only

Formula: 194.035 g/mol
MW: C₁₀H₁₅BO₃
Storage Temperature: Ambient
MDL Number: MFCD18384079
CAS Number: 1192107-39-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR