2-Bromo-4-chloropyridine-3-carboxylic acid ≥97%

Supplier: AOBChem USA

Warning

20527-5G 20527-1G 20527-25G
77634-334EA 196 USD
77634-334 77637-166 77654-130
2-Bromo-4-chloropyridine-3-carboxylic acid ≥97%
2-Bromo-4-chloropyridine-3-carboxylic acid

Precaución: For Research Use Only

Formula: 236.451 g/mol
MW: C₆H₃BrClNO₂
Storage Temperature: Ambient
MDL Number: MFCD18257846
CAS Number: 1256833-88-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR