3-Fluoro-2-methoxyisonicotinic acid ≥95%

Supplier: AOBChem USA

Warning

20495-5G 20495-1G 20495-25G 20495-500MG
77634-332EA 426 USD
77634-332 77637-164 77654-124 77645-168
3-Fluoro-2-methoxyisonicotinic acid ≥95%
3-Fluoro-2-methoxyisonicotinic acid

Precaución: For Research Use Only

Formula: C₇H₆FNO₃
Storage Temperature: Ambient
MDL Number: MFCD13185871
CAS Number: 1214362-24-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR