3-Chloro-2-phenoxypyridine ≥97%

Supplier: AOBChem USA

Warning

204363-5G 204363-1G 204363-25G 204363-500MG
77628-070EA 1599.59 USD
77628-070 77637-162 77654-120 77649-226
3-Chloro-2-phenoxypyridine ≥97%
3-Chloro-2-phenoxypyridine

Precaución: For Research Use Only

Formula: 205.64 g/mol
MW: C₁₁H₈ClNO
Storage Temperature: Ambient
MDL Number: MFCD29761080
CAS Number: 76893-57-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR