3-Bromo-5-methylbenzaldehyde oxime ≥97%

Supplier: AOBChem USA

Warning

20409-5G 20409-25G 20409-1G 20409-500MG
77634-328EA 313 USD
77634-328 77654-116 77641-176 77649-224
3-Bromo-5-methylbenzaldehyde oxime ≥97%
3-Bromo-5-methylbenzaldehyde oxime

Precaución: For Research Use Only

Formula: 214.059 g/mol
MW: C₈H₈BrNO
Storage Temperature: Ambient
MDL Number: MFCD19105368
CAS Number: 1205515-03-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR