Benzoic acid, 3-bromo-5-formyl-4-hydroxy-, methyl ester ≥97%

Supplier: AOBChem USA

Warning

203741-5G 203741-25G 203741-1G 203741-500MG
77634-320EA 591 USD
77634-320 77657-928 77641-166 77645-158
Benzoic acid, 3-bromo-5-formyl-4-hydroxy-, methyl ester ≥97%
Benzoic acid, 3-bromo-5-formyl-4-hydroxy-, methyl ester

Precaución: For Research Use Only

Formula: C₉H₇BrO₄
Storage Temperature: Ambient
MDL Number: MFCD22565644
CAS Number: 706820-79-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR