1-[2-(4-Chlorophenyl)ethynyl]cyclopentanol ≥95%

Supplier: AOBChem USA

Warning

203218-5G 203218-1G 203218-500MG
77628-068EA 1484.23 USD
77628-068 77641-164 77645-156
1-[2-(4-Chlorophenyl)ethynyl]cyclopentanol ≥95%
1-[2-(4-Chlorophenyl)ethynyl]cyclopentanol

Precaución: For Research Use Only

Formula: 220.7 g/mol
MW: C₁₃H₁₃ClO
Storage Temperature: Ambient
CAS Number: 1417462-82-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR