2,3-Difluorothioanisole ≥97%

Supplier: AOBChem USA

Warning

20314-5G 20314-1G 20314-25G 20314-500MG
77631-974EA 721 USD
77631-974 77637-156 77654-110 77645-154
2,3-Difluorothioanisole ≥97%
2,3-Difluorothioanisole

Precaución: For Research Use Only

Formula: C₇H₆F₂S
Storage Temperature: Ambient
MDL Number: MFCD13185574
CAS Number: 130922-38-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR