(5-Chloro-2,3-difluoropyridin-4-yl)(cyclohexyl)methanol ≥97%

Supplier: AOBChem USA

Warning

202944-5G 202944-1G 202944-25G 202944-500MG
77628-066EA 1807.65 USD
77628-066 77637-154 77654-106 77645-152
(5-Chloro-2,3-difluoropyridin-4-yl)(cyclohexyl)methanol ≥97%
(5-Chloro-2,3-difluoropyridin-4-yl)(cyclohexyl)methanol

Precaución: For Research Use Only

Formula: C₁₂H₁₄ClF₂NO
Storage Temperature: Ambient
MDL Number: MFCD34865607
CAS Number: 2969407-78-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR