(5-Bromo-2-fluoro-3-methylphenyl)(cyclohexyl)methanol ≥95%

Supplier: AOBChem USA

Warning

202860-5G 202860-1G 202860-500MG 202860-25G
77628-062EA 1583.11 USD
77628-062 77637-152 77652-458 77654-104
(5-Bromo-2-fluoro-3-methylphenyl)(cyclohexyl)methanol ≥95%
(5-Bromo-2-fluoro-3-methylphenyl)(cyclohexyl)methanol

Precaución: For Research Use Only

Formula: 301.195 g/mol
MW: C₁₄H₁₈BrFO
Storage Temperature: Ambient
MDL Number: MFCD34865523
CAS Number: 2969443-60-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR