4-Fluoro-2-(methoxymethyl)bromobenzene ≥95%

Supplier: AOBChem USA

Warning

20265-5G 20265-25G 20265-1G 20265-500MG
77631-970EA 618 USD
77631-970 77654-102 77641-156 77649-214
4-Fluoro-2-(methoxymethyl)bromobenzene ≥95%
4-Fluoro-2-(methoxymethyl)bromobenzene

Precaución: For Research Use Only

Formula: C₈H₈BrFO
Storage Temperature: Ambient
MDL Number: MFCD04116323
CAS Number: 842167-67-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR