4-Fluoro-2-(methoxymethyl)phenylboronic acid ≥95%

Supplier: AOBChem USA

Warning

20264-5G 20264-1G 20264-500MG
77628-060EA 1583.11 USD
77628-060 77641-154 77645-148
4-Fluoro-2-(methoxymethyl)phenylboronic acid ≥95%
4-Fluoro-2-(methoxymethyl)phenylboronic acid

Precaución: For Research Use Only

Formula: C₈H₁₀BFO₃
Storage Temperature: Ambient
MDL Number: MFCD28009343
CAS Number: 946607-86-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR