3-Phenoxyphenylboronic acid ≥95%
Supplier: AOBChem USA
|
Warning
|
Synonyms:
(3-Phenoxyphenyl)boronic acid, 3-Phenoxybenzeneboronic acid
20246-5G
20246-25G
77634-312EA
130
USD
77634-312
77657-924
3-Phenoxyphenylboronic acid ≥95%
3-Phenoxyphenylboronic acid
Precaución: For Research Use Only
Formula:
C₁₂H₁₁BO₃ MW: 214.03 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD07780272 CAS Number: 221006-66-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...