2-Bromo-1-(difluoromethoxy)-4-iodobenzene ≥97%

Supplier: AOBChem USA

Warning

202255-5G 202255-1G 202255-25G 202255-500MG
77631-968EA 1327 USD
77631-968 77637-150 77654-098 77645-146
2-Bromo-1-(difluoromethoxy)-4-iodobenzene ≥97%
2-Bromo-1-(difluoromethoxy)-4-iodobenzene

Precaución: For Research Use Only

Formula: 348.911 g/mol
MW: C₇H₄BrF₂IO
Storage Temperature: Ambient
MDL Number: MFCD18390924
CAS Number: 1062615-36-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR