3,5-Difluoro-4-iodopyridine ≥95%
Supplier: AOBChem USA
|
Warning
|
20207-5G
20207-1G
20207-25G
77634-308EA
179
USD
77634-308
77637-146
77657-920
3,5-Difluoro-4-iodopyridine ≥95%
3,5-Difluoro-4-iodopyridine
Precaución: For Research Use Only
Formula:
C₅H₂F₂IN Storage Temperature: Ambient |
MDL Number:
MFCD13185558 CAS Number: 1116099-07-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...