3,5-Dichloro-4-propoxybenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

20176-5G 20176-1G 20176-25G 20176-500MG
77631-966EA 1289 USD
77631-966 77637-144 77654-096 77645-142
3,5-Dichloro-4-propoxybenzaldehyde ≥97%
3,5-Dichloro-4-propoxybenzaldehyde

Precaución: For Research Use Only

Formula: 233.091 g/mol
MW: C₁₀H₁₀Cl₂O₂
Storage Temperature: Ambient
MDL Number: MFCD06246856
CAS Number: 883532-96-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR