4-Butoxy-3,5-dichloro-benzaldehyde ≥95%

Supplier: AOBChem USA

Warning

20174-5G 20174-25G 20174-1G 20174-500MG
77631-964EA 871 USD
77631-964 77657-918 77641-150 77649-210
4-Butoxy-3,5-dichloro-benzaldehyde ≥95%
4-Butoxy-3,5-dichloro-benzaldehyde

Precaución: For Research Use Only

Formula: 247.118 g/mol
MW: C₁₁H₁₂Cl₂O₂
Storage Temperature: Ambient
MDL Number: MFCD04129455
CAS Number: 50906-43-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR