(5-(Benzyloxy)-2-formylphenyl)boronic acid ≥95%

Supplier: AOBChem USA

Warning

20132-5G 20132-1G 20132-25G 20132-500MG
77634-306EA 433 USD
77634-306 77637-142 77657-916 77645-138
(5-(Benzyloxy)-2-formylphenyl)boronic acid ≥95%
(5-(Benzyloxy)-2-formylphenyl)boronic acid

Precaución: For Research Use Only

Formula: C₁₄H₁₃BO₄
MW: 256.06 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18383732
CAS Number: 1226773-36-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR