4-Chloro-2-fluoro-1-propoxybenzene ≥95%

Supplier: AOBChem USA

Warning

201235-5G 201235-1G 201235-25G 201235-500MG
77631-962EA 912 USD
77631-962 77637-140 77654-094 77649-208
4-Chloro-2-fluoro-1-propoxybenzene ≥95%
4-Chloro-2-fluoro-1-propoxybenzene

Precaución: For Research Use Only

Formula: 188.627 g/mol
MW: C₉H₁₀ClFO
Storage Temperature: Ambient
MDL Number: MFCD29044030
CAS Number: 1864734-30-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR