1-Fluoro-4-isobutoxybenzene ≥97%

Supplier: AOBChem USA

Warning

20047-5G 20047-25G 20047-1G 20047-500MG
77634-302EA 489 USD
77634-302 77657-912 77641-148 77649-202
1-Fluoro-4-isobutoxybenzene ≥97%
1-Fluoro-4-isobutoxybenzene

Precaución: For Research Use Only

Formula: C₁₀H₁₃FO
Storage Temperature: Ambient
MDL Number: MFCD19105366
CAS Number: 97295-04-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR