1-Iodo-2-methoxynaphthalene ≥95%

Supplier: AOBChem USA

Warning

20001-5G 20001-25G 20001-1G 20001-500MG
77634-300EA 552 USD
77634-300 77654-090 77641-144 77645-128
1-Iodo-2-methoxynaphthalene ≥95%
1-Iodo-2-methoxynaphthalene

Precaución: For Research Use Only

Formula: 284.093 g/mol
MW: C₁₁H₉IO
Storage Temperature: Ambient
MDL Number: MFCD00971813
CAS Number: 32721-21-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR