4-Bromo-2-ethylbenzoic acid ethyl ester ≥95%

Supplier: AOBChem USA

Warning

19984-5G 19984-1G 19984-500MG
77628-056EA 1583.11 USD
77628-056 77641-142 77649-200
4-Bromo-2-ethylbenzoic acid ethyl ester ≥95%
4-Bromo-2-ethylbenzoic acid ethyl ester

Precaución: For Research Use Only

Formula: C₁₁H₁₃BrO₂
Storage Temperature: Ambient
MDL Number: MFCD22490686
CAS Number: 1416979-60-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR