1-(4,6-Dichloropyridin-3-yl)ethan-1-one ≥95%

Supplier: AOBChem USA

Warning

Synonyms: 1-(4,6-Dichloropyridin-3-yl)ethanone, 5-Acetyl-2,4-dichloropyridine

199673-5G 199673-1G 199673-25G 199673-500MG
77631-956EA 685 USD
77631-956 77637-136 77654-086 77649-196
1-(4,6-Dichloropyridin-3-yl)ethan-1-one ≥95%
1-(4,6-Dichloropyridin-3-yl)ethan-1-one

Precaución: For Research Use Only

Formula: C₇H₅Cl₂NO
MW: 190.03 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03265322
CAS Number: 887573-44-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR