4-Fluoro-2-(1-methylethoxy)pyridine ≥97%

Supplier: AOBChem USA

Warning

199432-5G 199432-1G 199432-500MG
77654-082EA 2404 USD
77654-082 77641-140 77649-194
4-Fluoro-2-(1-methylethoxy)pyridine ≥97%
4-Fluoro-2-(1-methylethoxy)pyridine

Precaución: For Research Use Only

Formula: 155.17 g/mol
MW: C₈H₁₀FNO
Storage Temperature: Ambient
CAS Number: 1394983-09-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR