2,6-Dichloro-3,5-dimethylpyridine ≥95%

Supplier: AOBChem USA

Warning

199082-5G 199082-1G 199082-25G 199082-500MG
77628-054EA 1484.23 USD
77628-054 77637-132 77657-906 77645-124
2,6-Dichloro-3,5-dimethylpyridine ≥95%
2,6-Dichloro-3,5-dimethylpyridine

Precaución: For Research Use Only

Formula: 176.04 g/mol
MW: C₇H₇Cl₂N
Storage Temperature: Ambient
MDL Number: MFCD28145154
CAS Number: 125849-95-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR