4-Butoxy-3,5-dichlorophenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

19902-5G 19902-25G
77634-296EA 59 USD
77634-296 77657-902
4-Butoxy-3,5-dichlorophenylboronic acid ≥97%
4-Butoxy-3,5-dichlorophenylboronic acid

Precaución: For Research Use Only

Formula: C₁₀H₁₃BCl₂O₃
Storage Temperature: Ambient
MDL Number: MFCD15143476
CAS Number: 1218790-72-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR