2,6-Difluoro-4-methylbenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

19877-5G 19877-25G 19877-1G 19877-500MG
77631-952EA 755 USD
77631-952 77654-080 77641-136 77649-188
2,6-Difluoro-4-methylbenzaldehyde ≥97%
2,6-Difluoro-4-methylbenzaldehyde

Precaución: For Research Use Only

Formula: C₈H₆F₂O
MW: 156.129 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11847293
CAS Number: 1201597-22-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR