4,6-Difluorosalicylic acid ≥97%

Supplier: AOBChem USA

Warning

198691-5G 198691-1G 198691-25G 198691-500MG
77634-294EA 448 USD
77634-294 77637-130 77657-900 77649-186
4,6-Difluorosalicylic acid ≥97%
4,6-Difluorosalicylic acid

Precaución: For Research Use Only

Formula: C₇H₄F₂O₃
Storage Temperature: Ambient
MDL Number: MFCD06203215
CAS Number: 189283-54-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR