2-(4-Bromophenyl)-2-ethyl-1,3-dioxolane ≥97%

Supplier: AOBChem USA

Warning

19847-5G 19847-1G 19847-25G 19847-500MG
77628-052EA 1761.3 USD
77628-052 77637-128 77657-898 77645-122
2-(4-Bromophenyl)-2-ethyl-1,3-dioxolane ≥97%
2-(4-Bromophenyl)-2-ethyl-1,3-dioxolane

Precaución: For Research Use Only

Formula: 257.124 g/mol
MW: C₁₁H₁₃BrO₂
Storage Temperature: Ambient
MDL Number: MFCD34755755
CAS Number: 56255-86-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR