(3-Bromo-4,5-dichlorophenyl)methanol ≥95%

Supplier: AOBChem USA

Warning

197794-5G 197794-1G 197794-25G 197794-500MG
77631-946EA 1441 USD
77631-946 77637-124 77654-076 77649-182
(3-Bromo-4,5-dichlorophenyl)methanol ≥95%
(3-Bromo-4,5-dichlorophenyl)methanol

Precaución: For Research Use Only

Formula: 255.924 g/mol
MW: C₇H₅BrCl₂O
Storage Temperature: Ambient
MDL Number: MFCD28737836
CAS Number: 1805023-61-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR