Ethyl 3-bromo-4,5-dichlorobenzoate ≥97%

Supplier: AOBChem USA

Warning

197793-5G 197793-25G 197793-1G 197793-500MG
77634-292EA 314 USD
77634-292 77657-892 77641-130 77649-180
Ethyl 3-bromo-4,5-dichlorobenzoate ≥97%
Ethyl 3-bromo-4,5-dichlorobenzoate

Precaución: For Research Use Only

Formula: C₉H₇BrCl₂O₂
MW: 297.96 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11846096
CAS Number: 1160574-85-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR