4-Iodo-1-methyl-5-phenyl-1H-pyrazole ≥95%

Supplier: AOBChem USA

Warning

197773-1G 197773-5G 197773-500MG
77634-290EA 798 USD
77634-290 77657-890 77645-118
4-Iodo-1-methyl-5-phenyl-1H-pyrazole ≥95%
4-Iodo-1-methyl-5-phenyl-1H-pyrazole

Precaución: For Research Use Only

Formula: C₁₀H₉IN₂
Storage Temperature: Ambient
MDL Number: MFCD32189924
CAS Number: 1356997-80-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR