4',5'-Dichloro-2'-fluoroacetophenone ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 1-(4,5-Dichloro-2-fluorophenyl)ethan-1-one

197629-5G 197629-500MG 197629-25G 197629-1G
77634-288EA 316 USD
77634-288 77652-450 77657-888 77641-126
4',5'-Dichloro-2'-fluoroacetophenone ≥97%
4',5'-Dichloro-2'-fluoroacetophenone

Precaución: For Research Use Only

Formula: C₈H₅Cl₂FO
MW: 207.029 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18632530
CAS Number: 1286734-89-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR