1-(2-Fluoro-1,1'-biphenyl-4-yl)ethanone ≥95%

Supplier: AOBChem USA

Warning

Synonyms: 1-(2-Fluoro-[1,1'-biphenyl]-4-yl)ethanone, 4-Acetyl-2-fluorobiphenyl

197261-5G 197261-1G 197261-25G 197261-500MG
77631-940EA 650 USD
77631-940 77637-118 77657-880 77649-174
1-(2-Fluoro-1,1'-biphenyl-4-yl)ethanone ≥95%
1-(2-Fluoro-1,1'-biphenyl-4-yl)ethanone

Precaución: For Research Use Only

Formula: C₁₄H₁₁FO
MW: 214.24 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00127698
CAS Number: 42771-79-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR