1-((4-Methoxybenzyl)oxy)-3-nitrobenzene ≥97%

Supplier: AOBChem USA

Warning

196507-5G 196507-1G 196507-25G 196507-500MG
77631-938EA 791 USD
77631-938 77637-116 77654-068 77645-114
1-((4-Methoxybenzyl)oxy)-3-nitrobenzene ≥97%
1-((4-Methoxybenzyl)oxy)-3-nitrobenzene

Precaución: For Research Use Only

Formula: 259.26 g/mol
MW: C₁₄H₁₃NO₄
Storage Temperature: Ambient
MDL Number: MFCD00666642
CAS Number: 131770-84-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR