2-Bromo-1-methoxy-4-[(trifluoromethyl)thio]benzene ≥97%

Supplier: AOBChem USA

Warning

195996-5G 195996-1G 195996-500MG
77657-876EA 2113 USD
77657-876 77641-118 77649-168
2-Bromo-1-methoxy-4-[(trifluoromethyl)thio]benzene ≥97%
2-Bromo-1-methoxy-4-[(trifluoromethyl)thio]benzene

Precaución: For Research Use Only

Formula: 287.1 g/mol
MW: C₈H₆BrF₃OS
Storage Temperature: Ambient
MDL Number: MFCD29066659
CAS Number: 1357624-52-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR