2-(5-Iodo-2-methylphenyl)-1,3-dioxolane ≥95%

Supplier: AOBChem USA

Warning

19591-5G 19591-25G 19591-1G 19591-500MG
77631-936EA 763 USD
77631-936 77654-066 77641-116 77649-166
2-(5-Iodo-2-methylphenyl)-1,3-dioxolane ≥95%
2-(5-Iodo-2-methylphenyl)-1,3-dioxolane

Precaución: For Research Use Only

Formula: 290.098 g/mol
MW: C₁₀H₁₁IO₂
Storage Temperature: Ambient
MDL Number: MFCD34755751
CAS Number: 2764732-19-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR