6-(2,2,2-Trifluoroethoxy)-2-pyridinecarbonitrile ≥97%

Supplier: AOBChem USA

Warning

195420-5G 195420-25G 195420-1G 195420-500MG
77628-048EA 1690.23 USD
77628-048 77635-520 77641-112 77645-112
6-(2,2,2-Trifluoroethoxy)-2-pyridinecarbonitrile ≥97%
6-(2,2,2-Trifluoroethoxy)-2-pyridinecarbonitrile

Precaución: For Research Use Only

Formula: C₈H₅F₃N₂O
Storage Temperature: Ambient
MDL Number: MFCD18256065
CAS Number: 1256786-21-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR