2,3-Dichloro-1-(methylthio)-4-(trifluoromethyl)benzene ≥95%

Supplier: AOBChem USA

Warning

195273-5G 195273-500MG 195273-25G 195273-1G
77628-044EA 1797.35 USD
77628-044 77652-446 77654-058 77641-106
2,3-Dichloro-1-(methylthio)-4-(trifluoromethyl)benzene ≥95%
2,3-Dichloro-1-(methylthio)-4-(trifluoromethyl)benzene

Precaución: For Research Use Only

Formula: 261.09 g/mol
MW: C₈H₅Cl₂F₃S
Storage Temperature: Ambient
MDL Number: MFCD34863363
CAS Number: 2706202-50-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR