2,3-Dichloro-4-(trifluoromethyl)benzoic acid ≥95%

Supplier: AOBChem USA

Warning

195270-1G 195270-5G 195270-500MG
77637-114EA 593 USD
77637-114 77654-056 77649-158
2,3-Dichloro-4-(trifluoromethyl)benzoic acid ≥95%
2,3-Dichloro-4-(trifluoromethyl)benzoic acid

Precaución: For Research Use Only

Formula: 259.01 g/mol
MW: C₈H₃Cl₂F₃O₂
Storage Temperature: Ambient
MDL Number: MFCD28505055
CAS Number: 871254-69-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR