1-[2,3-Dichloro-4-(trifluoromethyl)phenyl]ethanone ≥95%
Supplier: AOBChem USA
|
Warning
|
195268-5G
195268-25G
195268-1G
195268-500MG
77628-042EA
1703.62
USD
77628-042
77654-054
77641-104
77649-156
1-[2,3-Dichloro-4-(trifluoromethyl)phenyl]ethanone ≥95%
1-[2,3-Dichloro-4-(trifluoromethyl)phenyl]ethanone
Precaución: For Research Use Only
Formula:
257.04 g/mol MW: C₉H₅Cl₂F₃O Storage Temperature: Ambient |
MDL Number:
MFCD28787599 CAS Number: 1804512-94-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...