Benzaldehyde, 2,3-dichloro-4-(trifluoromethyl)- ≥97%

Supplier: AOBChem USA

Warning

195261-5G 195261-1G 195261-500MG
77657-872EA 2186 USD
77657-872 77641-102 77649-154
Benzaldehyde, 2,3-dichloro-4-(trifluoromethyl)- ≥97%
Benzaldehyde, 2,3-dichloro-4-(trifluoromethyl)-

Precaución: For Research Use Only

Formula: 243.01 g/mol
MW: C₈H₃Cl₂F₃O
Storage Temperature: Ambient
MDL Number: MFCD31448313
CAS Number: 134099-42-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR