2,3-Dichloro-1-methyl-4-(trifluoromethyl)benzene ≥95%

Supplier: AOBChem USA

Warning

195252-5G 195252-25G 195252-1G 195252-500MG
77628-040EA 1561.48 USD
77628-040 77654-052 77641-100 77649-152
2,3-Dichloro-1-methyl-4-(trifluoromethyl)benzene ≥95%
2,3-Dichloro-1-methyl-4-(trifluoromethyl)benzene

Precaución: For Research Use Only

Formula: C₈H₅Cl₂F₃
Storage Temperature: Ambient
MDL Number: MFCD06658274
CAS Number: 115571-58-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR