4-Borono-3,5-dimethylbenzoic acid ≥97%

Supplier: AOBChem USA

Warning

19520-5G 19520-1G 19520-500MG
77628-038EA 1583.11 USD
77628-038 77637-112 77645-110
4-Borono-3,5-dimethylbenzoic acid ≥97%
4-Borono-3,5-dimethylbenzoic acid

Precaución: For Research Use Only

Formula: C₉H₁₁BO₄
Storage Temperature: Ambient
MDL Number: MFCD20659032
CAS Number: 851335-18-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR