2,6-Dichloro-3-hydroxy-4-methoxybenzaldehyde ≥95%

Supplier: AOBChem USA

Warning

194877-5G 194877-500MG 194877-25G 194877-1G
77628-036EA 1484.23 USD
77628-036 77652-444 77654-050 77641-098
2,6-Dichloro-3-hydroxy-4-methoxybenzaldehyde ≥95%
2,6-Dichloro-3-hydroxy-4-methoxybenzaldehyde

Precaución: For Research Use Only

Formula: C₈H₆Cl₂O₃
Storage Temperature: Ambient
MDL Number: MFCD04039898
CAS Number: 160431-96-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR