1-(2,6-Dichloro-4-methoxyphenyl)ethanone ≥95%
Supplier: AOBChem USA
|
Warning
|
194870-5G
194870-25G
194870-1G
194870-500MG
77631-932EA
969
USD
77631-932
77654-048
77641-096
77645-108
1-(2,6-Dichloro-4-methoxyphenyl)ethanone ≥95%
1-(2,6-Dichloro-4-methoxyphenyl)ethanone
Precaución: For Research Use Only
Formula:
219.06 g/mol MW: C₉H₈Cl₂O₂ Storage Temperature: Ambient |
MDL Number:
MFCD31631437 CAS Number: 157487-30-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...