1-Bromo-3-chloro-2-fluoro-4-(trifluoromethyl)benzene ≥97%

Supplier: AOBChem USA

Warning

194722-5G 194722-1G 194722-25G 194722-500MG
77628-034EA 1484.23 USD
77628-034 77637-110 77657-870 77649-148
1-Bromo-3-chloro-2-fluoro-4-(trifluoromethyl)benzene ≥97%
1-Bromo-3-chloro-2-fluoro-4-(trifluoromethyl)benzene

Precaución: For Research Use Only

Formula: 277.44 g/mol
MW: C₇H₂BrClF₄
Storage Temperature: Ambient
MDL Number: MFCD34863104
CAS Number: 2168156-96-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR