2-Chloro-3-fluorobenzotrifluoride ≥97%

Supplier: AOBChem USA

Warning

194645-5G 194645-1G 194645-25G
77634-278EA 231 USD
77634-278 77637-108 77654-046
2-Chloro-3-fluorobenzotrifluoride ≥97%
2-Chloro-3-fluorobenzotrifluoride

Precaución: For Research Use Only

Formula: C₇H₃ClF₄
MW: 198.55 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11100537
CAS Number: 1099597-97-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR